Would you like to log in again?
RAPID PURGE VALVE
Supplier: Chemglass
RAPID PURGE VALVE, ADJUSTABLE^ Rapid Purge Valve, Complete. Valve is designed for applications that require evacuated apparatus to be purged with a inert gas such as nitrogen, argon, etc.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
RAPID PURGE VALVE 80072-674 CG-4535-01 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty