Would you like to log in again?
Murphy and Read Spring, Stainless Steel Springs, Chemglass
Supplier: Chemglass
CG-110-03 CG-110-01
80072-944PK 43.12 USD InStock
80072-944 80072-948
Murphy and Read Spring, Stainless Steel Springs, Chemglass
Springs
Stainless steel springs are used to connect apparatus with hooks.
  • Springs can easily connect to almost any size hook
  • Diameter of the coils on all sizes is 0.190"
  • Stainless steel construction
All springs have an enlarged 1/4" I.D. loop at each end for easy connection to almost any size hook. Length listed is the distance between loops.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty