Would you like to log in again?
SOCKET JOINT 12/2
Supplier: Chemglass
SPHERICAL JOINT, SOCKET MEMBER^ Joint size: 12/2. Clamp Size: 12; Tube O.D; 7.5mm. Made to ASTM Specifications E677. For clamps, see CG-145-B and CG-150.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SOCKET JOINT 12/2 80073-092 CG-121-02 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty