Would you like to log in again?
SOCKET JOINT POLISHED 35/25
Supplier: Chemglass
SPHERICAL JOINT, POLISHED SOCKET MEMBER^ Joint size: 35/25. Clamp Size: 35; Tube O.D; 28mm. For use with CG-122, O-ring ball joint to provide for a flexible high vacuum seal. Polished sealing surface does not abrade o-ring. Note: Not for use with CG-120 ground ball joints.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SOCKET JOINT POLISHED 35/25 80073-130 CG-121-P-06 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty