Would you like to log in again?
OUTER JOINT #13 STOPPER NECK
Supplier: Chemglass
GROUND JOINT, FLASK LENGTH^ OUTER JOINTS. Tubing O.DxWall: 15x1.2mm. Joint size: 1/13. Standard taper flask length joints, both inner and outer member. Part number CG-125-09 is used as a replacement neck for 2 liter Squibb style separatory funnels.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
OUTER JOINT #13 STOPPER NECK 80073-192 CG-125-02 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty