Would you like to log in again?
JOINT DOUBLE SIDE SEAL 19/38
Supplier: Chemglass
SIDE SEAL JOINT^ Joint size: 19/38. Sidearm O.D; Two-10mm; Hole Size: 4mm. Outer joint having a tubulation sealed at the midpoint of the ground portion, and a corresponding hold drilled in the inner joint permitting venting of the side tube when the inner joint is turned to the proper postion.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
JOINT DOUBLE SIDE SEAL 19/38 80073-238 CG-132-03 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty