Would you like to log in again?
Gas Dispersion Tubes, Fritted, Chemglass
Supplier: Chemglass
CG-203-01 CG-203-03 CG-203-02 CG-203-04
80073-612EA 63.13 USD InStock
80073-612 80073-610 80073-616 80073-614
Gas Dispersion Tubes, Fritted, Chemglass
Gas Tubes
Tubes have a hollow fritted cylinder at the end of an 8 mm O.D. tube.
Cylinder dimensions: 120O.D.x20L mm (1/2x3/4"). Overall tube length: 254 mm (10").
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty