Would you like to log in again?
CHEM-THREAD COMPLETE # 12
Supplier: Chemglass
CHEM-THREAD, COMPLETE^ Size: 12. Tube O.D: 17mm; Accepts Tubing O.D: 9-12mm; O-Ring Size: 205. Consists of an externally threaded glass tube, Viton o-ring and Celcon compression cap. Tube O.D. listed is the shank O.D. of the connector.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CHEM-THREAD COMPLETE # 12 80073-968 CG-350-06 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty