Would you like to log in again?
Stopcocks, Straight Bore, Pressure, Glass Plugs, Chemglass
Supplier: Chemglass
CG-458-02 CG-458-01
80074-270EA 134.78 USD InStock
80074-270 80074-272
Stopcocks, Straight Bore, Pressure, Glass Plugs, Chemglass
Stopcocks
Solid glass plug has a threaded glass snelling which accepts an assembly similar to the one supplied with standard PTFE stopcocks.
This consists of a PTFE washer, PTFE nut, and Viton O-ring. Assembly provides for uniform seating tension to be applied to the plug which is precision ground and hand-lapped with corresponding numbers on the barrel and plug.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty