Would you like to log in again?
Stopcocks, High Vacuum, Straight Bore, Hollow Glass Plugs, Chemglass
Supplier: Chemglass
CG-471-01 CG-471-02
80074-306EA 207.72 USD InStock
80074-306 80074-308
Stopcocks, High Vacuum, Straight Bore, Hollow Glass Plugs, Chemglass
Stopcocks
Hollow plug, straight bore stopcock is precision ground and hand-lapped with corresponding numbers on barrel and plug.
Plug only part numbers are not supplied with retaining clip.
Delivery information: Supplied complete with the listed size CG-423 metal retaining clip.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty