Would you like to log in again?
PLUG ONLY VITON O'RINGS
Supplier: Chemglass
VALVE, NMR TUBE^ Replacement plug only, Viton o-rings. Valve is designed to be sealed to the top of either 5 or 10mm O.D. NMR tubes. Upper portion of the stem is extended with an additional o-ring for attachment to a piece of 7mm I.D.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PLUG ONLY VITON O'RINGS 80074-406 CG-512-04 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty