Would you like to log in again?
VALVE HI-VAC 1 ARM VITON 0
Supplier: Chemglass
VALVE, TFE, O-RING, HIGH VACUUM, 1-ARM^ Bore: 0-20mm; O-Ring Type: Viton. Valve has single sidearm sealed at a 90[degree] angle to the barrel.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VALVE HI-VAC 1 ARM VITON 0 80074-476 CG-534-04 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty