Would you like to log in again?
JOINT QUARTZ OUTER 19/38
Supplier: Chemglass
GROUND JOINT, OUTER, QUARTZ^ FULL LENGTH. Tubing O.D: 22.2mm. Joint size: 19/38. Clear fused quartz outer joints in medium and full length.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
JOINT QUARTZ OUTER 19/38 80075-212 CGQ-0132-14 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty