Would you like to log in again?
GrantBio Combi-spin Centrifuge/Vortex Mixer, Grant Instruments
Supplier: Grant
GrantBio
PCV-2400 USA
80076-360EA 1386.96 USD InStock
80076-360
GrantBio Combi-spin Centrifuge/Vortex Mixer, Grant Instruments
Shakers and Mixers Vortex Mixers
Cost-effective, fixed-speed, combined micro centrifuge/vortex mixer for combined or independent centrifuge and mixing applications of microtubes and 0.2ml microtube strips in low volume applications.
  • Centrifugation speed: fixed at 2800rpm/50Hz and 3500rpm/60Hz
  • Continuous operation or short spin
  • Choice of interchangeable rotors for different microtube sizes/combinations and for 0.2ml strips
  • Can be used at temperatures from +4ºC to +40ºC
Tubes are loaded into the rotor for simultaneous spinning, then removed for individual mixing in the vortex cup located at the top of the central shaft. In spin-mix-spin applications, the Combi-spin™ can be used with very low reagent volumes representing an overall saving in time, labour and material.
Certifications: CE marked.
Ordering information: Includes three-year manufacturer's warranty. Each unit comes complete with two interchangeable rotors: one holds twelve 1.5/2.0mL microtubes, the other holds twelve 0.5mL plus twelve 0.2mL microtubes. Additional rotors are available separately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty