Would you like to log in again?
E-C Sample Vials, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
W224810 W224809
80076-560CS 216.61 USD InStock
80076-560 80076-562
E-C Sample Vials, WHEATON®, DWK Life Sciences
Vials
Organic series clear glass vials are made from Wheaton 33® low extractable borosilicate glass.
Vials provide an inert and chemically-resistant environment for harsh, sensitive, and reactive samples. Vials accommodate size 24-400 screw caps (not included).
Certifications: Vials meet ASTM Type I Class A and USP Type I standards.
Ordering information: Packed in two shrink-wrapped packs of 100 vials.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty