Would you like to log in again?
Sample Vials, Borosilicate Glass, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
W224820 W224585 224982 W224840 224881 W224682 225290 224888 W224586 225288 224884 W224590 224885 224981 W224581 W224681 224882 224886 W224588 W224584 224984 W224589 W224684 W224604 W224605 W224582 W224591 225289
80086-692CS 179.08 USD InStock
80086-692 66011-405 14231-554 14231-556 14231-552 80086-690 66014-389 14230-836 14230-834 14230-832 80076-574 14230-830 80076-572 66011-937 66011-959 66011-358 66011-314 66011-336 66011-256 66011-278 66014-236 14230-828 66011-290 14230-826 14230-824 14230-822 14230-820 66011-948
Sample Vials, Borosilicate Glass, WHEATON®, DWK Life Sciences
Vials
These sample vials are made from low extractable borosilicate glass to provide superior chemical resistance.
Vials are available with or without caps and a choice of liners. PTFE faced rubber lined caps provide an inert and chemically-resistant barrier for reactive samples. Caps with 14B rubber liner are suitable for aqueous samples. Vials and caps are autoclavable.
Ordering information: Vials are packaged in Lab File® partitioned trays that provide an easy way to inventory samples or store empty vials. Caps are attached to vials to maintain cleanliness.
Clear vials conform to ASTM E438 Type 1, Class A and USP Type 1 standards. Amber vials conform to USP Type I requirements for light transmission to protect light-sensitive products.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty