Would you like to log in again?
Sample Vials, Borosilicate Glass, with Open-Top Screw Cap, Wheaton, DWK Life Sciences
Supplier: DWK Life Sciences
W224612 W224610 W224609 W224614
80076-580CS 267.2 USD InStock
80076-580 80076-578 80076-586 80076-584
Sample Vials, Borosilicate Glass, with Open-Top Screw Cap, Wheaton, DWK Life Sciences
Vials
Vials are made of Wheaton 33® clear or Wheaton 320 amber low extractable borosilicate glass.
Especially suited for research studies where stability of the glass is of prime importance. Vials are autoclavable. White polypropylene open-top caps have a specially designed septum of 5 mil of fluoropolymer resin facing 0.12 mil of silicone. The white polypropylene is glass-filled to provide extra chemical and heat resistance compared to standard polypropylene. The fluoropolymer resin-faced silicone septum is bonded to the cap, providing a permanent connection.
Certifications: Clear vials meet ASTM Type 1 Class A and USP Type 1 standards. Amber vials meet ASTM Type 1 Class B and USP Type 1 standards.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty