Would you like to log in again?
GLOVE STERILE SIZE 7.5 PK10PR
Supplier: Ansell Healthcare
DermaShield* Sterile Glove^ Made from a proprietary poly-chloroprene blend, free of latex proteins, and accelerators. Ideal choice in preventing both type I and type IV allergies, while still providing softness, and flexibility of latex gloves. Style 73-701, powder free, straight cuff. Size 7.5
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVE STERILE SIZE 7.5 PK10PR 80076-652 387071 Case of 200 Retrieving
Pack of 20 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty