Would you like to log in again?
DuPont™ Tyvek® IsoClean® Coveralls with Attached Hood
Supplier: DuPont
Tyvek® IsoClean®
IC180SWH2X002500 IC180SWH2X002500 IC180SWHXL002500 IC180SWH3X002500 IC180SWHXL002500 IC180SWH3X002500 IC180SWHMD002500 IC180SWHLG002500 IC180SWHLG002500
80077-172CS 386.95 USD InStock
80077-172 89129-624 CA80077-174 80077-176 CA80077-172 80077-174 CA80077-178 CA80077-176 80077-178
DuPont™ Tyvek® IsoClean® Coveralls with Attached Hood
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Serged seams
  • Attached elastic hood
  • Set sleeve design
  • Elastic wrists and ankles
  • Zipper closure
Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Coveralls have elastic openings for a tighter fit at wrists and ankles. Attached hood with elastic around face opening ensures a secure fit. Front zipper closure allows for easy donning and doffing.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty