Would you like to log in again?
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: Eppendorf
Eppendorf® Protein LoBind®
0030108302 022431081 0030108531 022431102 0030108531 022431081 0030108450 0030122356 0030122283 0030122283 0030122240 0030122216 022431102 022431064 022431064 0030108302
76683-144PK 166.18 USD InStock
76683-144 CA76683-150 76683-150 76289-498 89429-314 76289-496 CA80077-232 80077-232 CA89429-314 CA76683-144 CA80077-226 76313-966 MSPP-030108450 80077-228 80077-226 CA80077-228
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Tubes Microcentrifuge Tubes
Engineered to minimize sample binding and elevate recovery, Eppendorf® Protein LoBind® tubes safeguard valuable proteins during processing. The cutting-edge LoBind® technology ensures optimal yields for Western blotting, mass spectrometry, and protein purification applications.
  • Surface-coating-free, including substances like silicone, to reduce the risk of sample interference
  • Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase, RNase, and PCR inhibitors
  • Available in tube, microplate, and deep-well plate formats for convenient upscaling of experiments
  • Snap, screw, and SnapTec® cap options available
Unlock the benefits of Eppendorf® Protein LoBind® tubes, which are meticulously designed to offer unparalleled advantages. Available in a range of sizes and formats, these tubes are precision-crafted to minimize sample binding, providing a protective shield for valuable proteins, particularly in low-concentration applications. The LoBind® technology not only diminishes the risk of sample loss but also elevates protein recovery, guaranteeing users extract the maximum yield from every precious sample. Gain peace of mind knowing that proteins are safeguarded and results are optimized with Eppendorf® Protein LoBind® Tubes.
Applications include preparation or storage of protein, peptide, or antibody samples, Enzymatic assays (the hydrophilic surface reduces denaturation when it comes into contact with the tube inner wall), prevention of sample loss during storage of virus stock solutions, and storage of cell suspensions.
Manufactured without the use of slip agents, plasticizers, and biocides - substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty