Would you like to log in again?
DuPont™ ProShield® 30 Boot Covers
Supplier: DuPont
ProShield®
PE444SWHXL010000 PE444SWHXL010000 PE444SWHLG010000 PE444SWHLG010000
80077-260CS 135.01 USD InStock
80077-260 CA80077-260 CA89129-708 89129-708
DuPont™ ProShield® 30 Boot Covers
Disposable Garments
ProShield® 30 shoe and boot covers offer exceptional skid resistance.
  • Serged seams
  • Elastic at ankles
  • Skid-resistant
ProShield® 30 is made from spun bonded polypropylene with a polyethylene coating. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Elastic at ankles ensures a secure fit.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty