Would you like to log in again?
Pulse Platform Mixer, Glas-Col®
Supplier: Glas-Col
099A PPM12
80078-694EA 9967.22 USD InStock
80078-694
Pulse Platform Mixer, Glas-Col®
Shakers and Mixers Vortex Mixers
Unit accomodates a wide variety of glassware, racks, well plates, or reactor blocks and generates precise vortexing action for accurate, repeatable results.
  • Touch screen interface
The microprocessor-based control features speed display; a programmable, multifunction timer; and selectable pulse modes. The unit has chemical-resistant foam on the top and driveplate and an RS-232 communications port.
Certifications: UL listed.
Ordering information: Supplied with a 1.5 m (5'), three-wire grounded cord.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty