Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dry Block Heater, Digital, 240 V Glas-Col®

Supplier: Glas-Col

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

R34563 099AHBC24 R34 561 R34 562 R34516
80078-742EA 442.15 USD InStock
80078-742 80078-744 80078-746 80078-748 80078-740
Dry Block Heater, Digital, 240 V Glas-Col®
Block Heaters Digital Block Heaters
This small, constant-temperature heater can accommodate a variety of different size blocks

  • 240 V

Maintains temperature from slightly above ambient to 125°C (257°F), with ±1°C accuracy. The heater opening is in standard SBS format, which allows heating of high- or low-profile standard well plates. The heater features single-loop, auto-tuning digital control with simple input commands, dual four-digit LED display for temperature in °C or °F, and up and down keys for set point adjustment. The heater also features PID control parameters that set automatically for optimum system performance, an on/off power switch, and fuse protection. The unit exterior is finished in foxtip gray and charcoal.

Ordering information: Heater includes a three-wire cord with molded plug.

Caution: c

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty