Would you like to log in again?
WHEATON® E-Z Crimper™ Hand-Operated Crimpers, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
E-Z Crimper™
W225300 W225303 W225302 W225301
80078-994EA 603.95 USD InStock
80078-994 80078-996 80078-990 80078-992
WHEATON® E-Z Crimper™ Hand-Operated Crimpers, DWK Life Sciences
Crimpers and Decappers
Crimpers feature polished crimping jaws for consistent sealing, a redesigned spring assembly, and a short, cushioned ergonomic handle that helps reduce hand fatigue and adds comfort
Note: Autoclaving may change the appearance or reduce product life.
An adjustable stop allows tightening of the seal while maintaining consistent crimping. The 8, 11, and 13mm models feature tapered jaws for easier vial access in rack. Each crimper size is labeled for quick identification. Autoclavable at 121°C (250°F).
Ordering information: Crimpers work with the Crimpmaster® crimping station. Crimper 80078-992 is supplied with an O-ring to provide the option of crimping the top of a vial to create an extra tight seal. This option is recommended only for standard-opening vials.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty