Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

WHEATON® E-Z Decappers™ Hand-Operated Decappers, DWK Life Sciences

Supplier: DWK Life Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

E-Z Decappers™
W225351 W225350 W225352 W225353
80079-002EA 747.53 USD InStock
80079-002 80079-000 80078-998 80079-004
WHEATON® E-Z Decappers™ Hand-Operated Decappers, DWK Life Sciences
Crimpers and Decappers
These durable, hand-operated decappers are an economical way to remove seals quickly and easily.

Note: Autoclaving may change the appearance or reduce product life.

To remove a seal, just place the decapper on the seal and squeeze the handles together. Decappers feature a cushioned ergonomic handle that helps reduce hand fatigue and adds comfort. The 8, 11, and 13mm models feature tapered jaws for easier vial access in racks. Each decapper size is labeled for quick identification. Autoclavable at 121°C (250°F).

Ordering information: Decappers work with the Crimpmaster® crimping station.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty