Would you like to log in again?
KoolTemp® Insulated Envelope, Cold Chain Technologies
Supplier: Cold Chain Technologies
KoolTemp®
IE1216-V
80079-854CS 1291.9 USD InStock
80079-854
KoolTemp® Insulated Envelope, Cold Chain Technologies
Envelopes
KoolTemp® insulated envelopes are lightweight, freight saving, durable, and designed with a spacious payload chamber for temperature-sensitive needs.
  • Lightweight, precision-cut expanded insulation
  • Spacious, crush-resistant, virtually airtight chamber
  • Ideal for personal transportation of medical samples, food, perishables or valuables
  • Seal to seal dimensions: 12×16"
An insulated envelope addresses the challenge of heat loss with efficient insulation, reflective exterior surface and a virtually airtight payload chamber.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty