Would you like to log in again?
Dippas™, Sample Dippers, Sterile, Dynalon
Supplier: Dynalon
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
507054-0100
80079-938CS 616.99 USD InStock
80079-938
Dippas™, Sample Dippers, Sterile, Dynalon
Samplers Sample Dippers
Sterile dippas enable samples of liquid or powder to be collected and sent for testing in the same container, eliminating the risk of cross contamination
Once the sample has been taken, the handle can be neatly snapped off. All dippas come individually wrapped and are guaranteed sterile by gamma irradiation.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty