Would you like to log in again?
BRAND Test Tube Racks, BrandTech
Supplier: BrandTech
4340022 4340012 4341050 4340020 4340000 4340021 4340001 4340040 4340061 4340010 4340062 4340030 4340032 4340011 4341051 4341001 4340031 4340060 4341003
89400-726PK 196.17 USD InStock
89400-726 89400-728 80080-342 80080-320 80080-310 80080-322 80080-312 80080-334 80080-330 80080-328 80080-318 89400-730 80080-302 80080-314 89400-732 80080-336 80080-326 80080-304 80080-338
BRAND Test Tube Racks, BrandTech
Racks Tube Racks
Sturdy polypropylene racks snap together for easy assembly.
  • Filled PP, density 1.2 g/cm³ so will not float in waterbath
  • Operating temperature -20 to 90 °C
  • Autoclavable (121 °C), (2 bar), acc. DIN EN 285
Versatile and inexpensive racks for common diameter test tubes. Alphanumerical positions for clear sample identification. Wide spaces between positions. The racks are delivered dismantled and can be put together securely and permanently in a few steps, without tools.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty