Would you like to log in again?
Polywipe-C Heatseal Wipes, Contec®
Supplier: Contec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HexaWipe™
PCHS-99 LWCS0033 PCHS-1212 PCHS-1418/100 PCHS-1418/100 LWCS0004 PCHS-99B/750 LWCS0024 LWCS0027 PCHS-1212B/150
76326-008CS 557.11 USD InStock
76326-008 80080-454 89368-630 77776-712 94024-466 76471-010 CA76471-010 68888-301 33500-052 10171-160
Polywipe-C Heatseal Wipes, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Polywipe-C heatseal wipes are made from a standardweight 100% knitted polyester that is laser cut to bond the fibers at the edges of the wipe.
  • Soft textured wipe cleans surfaces without scratching
  • Good sorbency with solvents
  • Available validated sterile
The wipe is chemical resistant and exceptionally low in particles and extractable residue making it ideal for critical cleaning. The wipes are laundered and packed in an ISO Class 4 cleanroom.
Certifications: Recommended for ISO Class 3 to 5 environments.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty