Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Barnstead/Lab-Line Unwire™ Test Tube/Microcentrifuge Tube Racks for Universal Platforms, Thermo Scientific
Supplier: Thermo Scientific
Lab-Line Unwire™
30182 30186 30181 30189 30184 30180BI 30190 30187 30191 30185 30183 30188
80080-562EA 546.09 USD InStock
80080-562 80080-552 80080-574 80080-564 80080-554 80080-570 80080-560 80080-572 80080-566 80080-556 80080-568 80080-558
Barnstead/Lab-Line Unwire™ Test Tube/Microcentrifuge Tube Racks for Universal Platforms, Thermo Scientific
Shakers and Mixers Shaking Incubators
Designed for use with universal platforms (see 57018-990 series)
ResMer® resin construction withstands extreme conditions, including autoclaving, mechanical washing, chemical disinfection, freezing, and temperatures up to 135 °C (275 °F). Racks are chemically resistant to low concentrations of acids and hydrocarbons, bases, alcohols, esters, and aldehydes.
Ordering information: Racks include a removable polyethylene foam insert to hold tubes tightly and prevent spinning.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty