Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whatman™ Quantitative Filter Papers, Ashless Grades (Ash 0.007%), Grade 40

Supplier: Whatman products (Cytiva)
Whatman™
1440-070 1440-329 1440-070 10380005 10380006 1440-329 10380004 1440-012 10380004 1440-6168 1440-917 1440-185 1440-320 1440-110 1440-090 1440-240 1440-055 1440-125 1440-042 1440-047 1440-150 1440-032 1440-185 1440-150 10380006 1440-917 1440-240 1440-032 1440-055 1440-125 1440-110 10380005 1440-090 1440-012 1440-042 1440-6168 1440-320 1440-047
89203-510PK 38.14 USD InStock
89203-510 CA11013-630 CA10147-982 28475-128 28475-106 CA11013-350 CA28475-015 28475-026 28475-004 28475-048 11008-520 70240-042 CA76292-862 CA11008-520 28475-140 28475-081 CA76292-860 28475-060 CA80080-586 76292-858 10035-258 10147-982 CA70240-042 CA89203-510 CA28475-128 CA28475-106 CA28475-026 CA28475-048 CA28475-004 28475-015 CA28475-140 80080-586 76292-860 CA28475-081 CA28475-060 76292-862 10035-260 CA76292-858
Whatman™ Quantitative Filter Papers, Ashless Grades (Ash 0.007%), Grade 40
Filters Filter Paper Quantitative Filter Paper
Whatman Grade 40 high-purity filters from Cytiva's business are general-purpose papers for quantitative analysis. Grade 40 filters are suitable for a variety of analytical filtration applications.

  • High-purity filters contain 0.007% maximum ash content
  • Medium nominal particle retention rating of 8 µm
  • Medium filtration speed (Herzberg 340 s)
  • Particularly useful for gravimetric analysis

High-purity filters support accurate analysis in applications sensitive to very low levels of background contamination and in gravimetric analysis where ash content might affect weight measurements of combusted samples.

Cytiva's high-purity Whatman Grade 40 ashless paper is suited to:
- Soil analysis, when separating solid matter from aqueous extracts
- Quantitative determination of milk sediment
- Use as a clean-up filter for solutions prior to atomic absorption (AA) spectrometry
- Analysis of constituent components in cements, clays, iron, and steel products
- Collection of trace elements and radionuclides from the atmosphere

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty