Would you like to log in again?
VWR® Odor Eliminators
Supplier: VWR
VWR®
04393-65 04393-61 04393-68 04393-62
80080-890CS 833.31 USD InStock
80080-890 80080-896 80080-898 80080-888
VWR® Odor Eliminators
Deodorants and Air Fresheners
These sterilizer deodorizers chemically absorb odor-causing molecules, removing unwanted odors from your facility rather than masking them.
  • Chemically absorbent deodorizers
Simply place one capsule in your autoclave or incubator with each load. The heat and moisture will dissolve the capsule and release a clean fresh scent.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty