Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Corning® Storage Mats, Corning

Supplier: Corning
Corning®
3341 3083 3346 3080
29445-122CS 464.55 USD InStock
29445-122 80081-124 80081-114 33500-570
Corning® Storage Mats, Corning
Seals Microplate Seals
Multiple formats for specific and precise fit on 96-well and 384-well plates and blocks.

  • Non sterile
  • Not treated

Round well microplate storage mat: Fits Corning® 96 round well plates and blocks.
Manufactured from ethyl vinyl acetate (EVA), compatible with DMSO
Certified free from DNase and RNase
Can be applied manually or with applicator

Square well storage block sealing mat: Fits Corning® 96 square 2 ml well storage blocks.
Manufactured from ethyl vinyl acetate (EVA), compatible with DMSO
Certified free from DNase and RNase
Can be applied manually or with applicator

Chemical resistant sealing mats: Precise fit for Corning® 384-well deep well PP storage block or Corning® 96-well expanded volume PP microplate.
Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g. DMSO, ethanol, methanol)
Firm seal prevents evaporation and gas exchange
Ideal for use in long-term, low temperature compound storage

Thermowell® sealing mat: These silicone rubber mats offer a cost effective alternative to other sealing methods and provide 100% sealing when used in conjunction with clamp or screw down heated lid thermal cyclers.
Easy to apply and remove
Fully autoclavable and reusable (at least five times)

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty