Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
KoolTemp® Insulated Shippers, EPS Foam Shipping Coolers, Cold Chain Technologies
Supplier: Cold Chain Technologies
KoolTemp®
TL888-SU-V TL8612-SU-V TL1500-SU-V TL8612-M-V TL644-M-V TL869-M-V TL12107-SU-V TL2000-M-V TL12109-SU-V TL3800-SU-V TL645-SU-V TL2800-SU-V TL867-SU-V TL2000-SU-V TL869-SU-V TL868-M-V TL3300-SU-V TL864-SU-V TL1189-SU-V TL1187-SU-V TL2300-SU-V
80081-212CS 158.58 USD InStock
80081-212 80081-210 33500-406 80081-204 80081-216 80081-202 33500-404 80081-214 80081-208 80081-206 33500-376 33500-410 33500-388 33500-396 33500-378 33500-400 33500-412 33500-390 33500-380 33500-394 33500-384
KoolTemp® Insulated Shippers, EPS Foam Shipping Coolers, Cold Chain Technologies
Mailers and Shippers
Lightweight and reusable insulated expanded polystyrene (Styrofoam) shippers provide excellent thermal protection for perishables and samples.
  • Reusable
  • Excellent thermal protection
Channel construction and stand-off pads keep product away from condensation and provide more uniform temperature control throughout the cooler when used with dry ice or Koolit® refrigerants. State-of-the-art lid interlock design and secure toungue-and-groove friction fit minimize thermal loss. Rounded corners resist friction damage.
Delivery information: Shippers are available with or without outer corrugated carton.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty