Would you like to log in again?
String Knit Gloves, MCR Safety
Supplier: MCR Safety
9507SM 9633XLM 9613SM 9630SM 9660SM 9630LM 9660LM 9660SMB
80081-662PK 26.51 USD InStock
80081-662 80081-660 89135-732 80081-664 80081-654 89135-736 89135-734 80081-658
String Knit Gloves, MCR Safety
Gloves
Our multi-purpose string knit gloves come in various blends and weights.
  • 7 gauge heavy weight gray cotton/polyester
  • Reversible pattern
  • Color coded hem to identify sizes
  • Made with FDA accepted materials
Economy, regular, and heavy weight gauge offerings are available. These gloves are suitable for general hand protection in light duty applications.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty