Would you like to log in again?
Memphis UltraTech® Gloves, Air Infused, MCR Safety
Supplier: MCR Safety
Memphis UltraTech®
9694XS 9694M 9694XL 9694S 9694L
80081-728PK 115.26 USD InStock
80081-728 80081-732 80081-730 80081-736 80081-734
Memphis UltraTech® Gloves, Air Infused, MCR Safety
Gloves
UltraTech® provides ultra comfort, ultra fit, and ultra protection.
  • The seamless nylon shell offers outstanding breathablility and comfort
  • The polyurethane palm and finger coating provides grip,puncture resistance, abrasion resistance and dexterity
Great for small parts handling. The 9694 features a 15 gauge white nylon shell and gray air infused nitrile coating on the palm and fingertips.
Ordering information: These gloves are polybagged by the pair.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty