Would you like to log in again?
Crews® Blackjack® Protective Eyewear, MCR Safety
Supplier: MCR Safety
Crews®
S2212 S2210AF S2219
80082-001PK 60.77 USD InStock
80082-001 80082-003 80081-999
Crews® Blackjack® Protective Eyewear, MCR Safety
Safety Glasses
These glasses feature a single, dual-focus wraparound lens design.
  • Integral side shields for added protection
  • Metal temples with spring hinge design provide maximum flexibility
Strong, lightweight, polycarbonate lenses with Duramass® scratch-resistant coating filter out 99% of UV radiation.
Certifications: Meets or exceeds ANSI Z87.1+ standards
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty