Would you like to log in again?
X-plore® Air-Purifying Respirators, Draeger
Supplier: Dräger
X-plore®
R55351 R55330 6738034 6738039 6738027 6738035 6738021 6738036 6738035 R55270 R55352 R55331 R55352 6738027 6738024 R55350 6738020 6738034 6738024 R55332 6738025 R55350 R55351 6738037 R55270 6738021 R55331 6738020 R55332 6738026 6738037 6738025
80082-482EA 13.01 USD InStock
CA80082-504 CA80082-506 CA80082-508 80082-482 CA80082-490 80082-484 CA80082-492 80082-486 80082-488 CA80082-496 80082-500 CA80082-498 CA80082-476 13453-582 80082-502 CA80082-478 80082-504 80082-506 80082-508 80082-490 CA80082-480 80082-492 CA80082-482 80082-494 CA80082-484 80082-496 CA80082-486 80082-498 80082-476 CA80082-488 80082-478 CA80082-500
X-plore® Air-Purifying Respirators, Draeger
Respirators Reusable Respirators Half Face Respirators
Respirators provide comfortable protection during industrial tasks, such as welding, painting, or chemical handling.
  • All masks use a two-point bayonet connection for safe and easy filter attachment
The 5500 series full-face masks offer complete respiratory and eye protection
The 3500 series reusable half-masks employ unique Draeger-Flex™ hypoallergenic material that has a soft and comfortable seal without skin irritation. The FlexiFit® harness system is extremely comfortable even under a hard hat. These compact, low-profile masks can be easily worn under face shields. The slide-down system allows mask removal without taking off a hard hat or face shield. A flexible nose seal provides a superior fit, even when wearing safety glasses. Innovative X-guided strap system ensures an even distribution of the mask seal against the face without unwanted pressure.
The 3300 series limited-use half-masks are constructed of a soft, durable TPE elastomer and have all of the features of the 3500 series reusable masks without the slide-down feature.
The large lens provides an excellent field of vision with a high level of chemical and mechanical resistance. A double-layer face seal and exclusive triple-seal edge allow anyone to wear the same size mask. The five-point head harness is easy to don and adjust.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty