Would you like to log in again?
Combined Ion Selective Electrodes with S7 Plug-In, WTW
Supplier: WTW
106395
80085-244EA 2906.07 USD InStock
80085-244
Combined Ion Selective Electrodes with S7 Plug-In, WTW
Electrodes ISE Electrodes
Gas sensitive combined electrode with S7 plug-in connection for ammonia/ammonium measurement in laboratories and in the field.
Delivery information: Supplied with protective cap, storing cap, three membrane caps, one bottle NH? electrolyte solution (50 ml) and operating manual.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty