Would you like to log in again?
Axygen® MaxyClear Snaplock Microcentrifuge Tube, Polypropylene, Spectrum, Non sterile, Corning
Supplier: Corning
Axygen®
MCT-060-SP MCT-150-SP
80086-022CS 949.13 USD InStock
80086-022 80086-024
Axygen® MaxyClear Snaplock Microcentrifuge Tube, Polypropylene, Spectrum, Non sterile, Corning
Tubes Microcentrifuge Tubes
Axygen MaxyClear microcentrifuge tubes are available in a variety of colors. Axygen Maxymum Recovery® surface and sterile options are also available.
  • 0,6, 1,5, and 2,0 ml clear microcentrifuge tubes are boil proof
  • Colored tubes are not boil-proof
  • Co-polymer tubes offer exceptional strength and chemical resistance
  • Clear microcentrifuge tubes are boil-proof
  • Maximum G-force is 14000 min⁻¹
  • RNase-/DNase-free and nonpyrogenic
  • Sterility Assurance Level (SAL) of 10⁻⁶
SnapLock microcentrifuge tubes have frosted cap surfaces and frosted side panels, ideal for labeling. Each cap features a piercing port in the center of the cap. Microcentrifuge tubes with no caps are available for some sizes.
Color - Assorted: Each pack contains a mixture of blue, green, violet, orange, red, and yellow.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty