Would you like to log in again?
Axygen® Pipette Tip 1200 µl with Graduation Marks, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Axygen®
TE-1004-B-L-R-S TE-1004-B-L-R TE-1004-B
80086-116CS 428.36 USD InStock
80086-116 80086-118 80086-108
Axygen® Pipette Tip 1200 µl with Graduation Marks, Corning
Pipette Tips
Axygen brand tips are designed to fit Eppendorf-Style 1000 µl single-channel pipettors.
Flexible walls and a series of internal sealing rings ensure a secure fit and less force required to load and eject. Accuracy and precision testing is performed for every manufacturing lot using the finest pipettors in the industry. These 1000 µl tip have graduation marks at 110 and 500 µl.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty