Would you like to log in again?
VACUUBRAND® Service Kit for MZ2C Chemistry-Design Pump, BrandTech
Supplier: BrandTech
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VACUUBRAND®
20696814
80086-246EA 819.21 USD InStock
80086-246
VACUUBRAND® Service Kit for MZ2C Chemistry-Design Pump, BrandTech
Services
Set of two diaphragms and four valves.
Does not fit "NT" models.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty