Would you like to log in again?
ASO Latex Test Kits, ASI
Supplier: Arlington Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ASI™
3001000 300025 300050 300100
80087-270EA 197.54 USD InStock
80087-270 80087-268 80087-266 80087-264
ASO Latex Test Kits, ASI
Clinical Diagnostic Supplies Clinical Reagents and Kits
  • 2 Minute slide test
  • 2 Liquid controls included
  • Longest shelf life - up to 18 month
  • Proven by AAB, CAP, and API surveys
The ASI ASO Latex Slide Test is a convenient and specific latex slide agglutination assay for the qualitative and semiquantitative detection of Antistreptolysin-O (ASO) antibodies in human serum. No initial dilution of patient sample is required for this test.
Certifications: CE Marked. CPT Code 86060. 510(k)-K853857.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty