Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

RPR Syphilis Test Kits, ASI

Supplier: Arlington Scientific
ASI™
90010000-1 900025 9005000-10 90010000-3 9005000-30 900500 900100
80087-338EA 5515.98 USD InStock
80087-338 80087-348 80087-336 80087-346 80087-344 80087-342 80087-340
RPR Syphilis Test Kits, ASI
Clinical Diagnostic Supplies Clinical Reagents and Kits
The ASI RPR Card Test is a screening test for the sexually transmitted disease, SYPHILIS. The disease is found worldwide and may be fatal if left untreated. To keep the blood supply safe, transfusion blood is tested for syphilis, HIV and hepatitis.

  • A CDC recommended primary screening test
  • Simple and economical to use
  • Easy to interpret
  • All testing materials provided
  • Sold with controls or without
  • 3 levels of liquid controls included: reactive, weak reactive, nonreactive
  • Color-coded dropper vials
  • Warp resistant coated test cards
  • Screw cap vials - No glass ampules
  • CPD, CPDA-1, EDTA, Heparin or Sodium Citrate may be used
  • Made with pride in the USA
  • Longest shelf life - up to 24 months
  • Proven results on CAP, AAB, and API Surveys
  • 10 and 30 well test cards available
  • Sensitivity : >98%
  • Specificity: >99%

Certifications: CE marked. CPT Code 86592. 510(k)-K851504

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty