Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VDRL Syphilis Test Kit, Arlington Scientific

Supplier: Arlington Scientific
ASI™
950010
80087-360EA 382.9 USD InStock
80087-360
VDRL Syphilis Test Kit, Arlington Scientific
Clinical Diagnostic Supplies Clinical Reagents and Kits
  • Suitable for testing cerebral spinal fluid (CSF) specimens
  • Meets CDC primary algorithm recommendation
  • Easy to interpret
  • Simple and economical to use
  • Screw cap vials - No glass ampules
  • Room temperature storage
  • Longest shelf life - up to 60 months
  • Proudly made in USA
  • Results on CAP, AAB and API surveys
  • Sensitivity & Specificity
  • Sensitivity: >99%
  • Specificity: >99%

The ASI VDRL Antigen test for syphilis is a specific qualitative and semi-quantitative nontreponemal flocculation test for the detection of reagin antibodies in human serum. The procedure is based on the VDRL antigen being combined at a correct ratio with buffered saline and then mixed with heat-inactivated serum.

Certifications: CE marked. CPT Code 86592.

Ordering information: Kits include two vials of VDRL antigen (2 x 5 mL), two vials of VDRL buffered saline (2 x 60 mL). The kit could perform as many as 5,900 tests but it is generally less due to facility testing workflow.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty