Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

C-Reactive Protein (CRP) Slide Test Kits, ASI

Supplier: Arlington Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

ASI™
5001000 500025 500100 500050
80087-370EA 1558.26 USD InStock
80087-370 80087-368 80087-374 80087-372
C-Reactive Protein (CRP) Slide Test Kits, ASI
Clinical Diagnostic Supplies Clinical Reagents and Kits
The ASI CRP Slide Test is a convenient and specific slide agglutination assay for the qualitative and semiquantitative detection of C-Reactive Protein (CRP) in human serum. No initial dilution of patient sample is required for this test.

  • 2 Minute slide test
  • No initial dilution required
  • 2 Liquid controls included
  • Longest dating - up to 24 month shelf life
  • Proudly made in USA
  • Proven by AAB, CAP and API surveys
  • Sensitivity: 6 mg/L
  • Specificity: >95%

Certifications: CE marked. CPT Code 86140. 510(k)-K821388

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty