Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SLE Latex Test Kits, ASI, Arlington Scientific

Supplier: Arlington Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

ASI™
350025 3501000 350050 350100
80087-392EA 2957.91 USD InStock
80087-392 80087-390 80087-396 80087-394
SLE Latex Test Kits, ASI, Arlington Scientific
Clinical Diagnostic Supplies Clinical Reagents and Kits
  • Proven reliability
  • Accurate screening test for SLE
  • 1 Minute test
  • 2 Liquid controls included
  • Proudly made in USA
  • Longest shelf life - up to 24 months
  • Sensitivity: >86%
  • Specificity: >82%

The ASI SLE Slide Test is a convenient and specific latex slide agglutination assay for the qualitative and semiquantitative detection of anti-deoxyribonucleoprotein (anti-DNP) in human serum. No initial dilution of patient sample is required for this test.

Certifications: CE marked. CPT Code 86225. 510(k)-K821385

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty