Would you like to log in again?
SP Wilmad-LabGlass Solvent Jet Washers for NMR Tubes, Economy, SP Industries
Supplier: SP Industries
WG-1207-J10 WG-1207-J5
80087-514EA 292.94 USD InStock
80087-514 80087-512
SP Wilmad-LabGlass Solvent Jet Washers for NMR Tubes, Economy, SP Industries
Tubes NMR Tubes
Reliable for over 40 years, this simple design washing unit is the most economical for single tube cleaning.
  • Each unit only accommodates NMR tube with Ext.Ø and length as indicated in specifications
  • Requires filter flask (sold separately; 500 ml recommended)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty