Would you like to log in again?
Axygen® AxyGem™ 96-Well Sitting-Drop Crystallography Plate, Corning
Supplier: Corning
Axygen®
CP-AXYGEM-96-50
80089-668CS 493.1 USD InStock
80089-668
Axygen® AxyGem™ 96-Well Sitting-Drop Crystallography Plate, Corning
Microplates HTS Microplates
SBS standard, sitting-drop, high throughput protein crystallization plate has 96 buffer wells, each with one corresponding protein well
The D-shaped buffer wells have a maximum volume of 250µL. The elevated protein wells have a maximum volume of 4.3µL and feature a round shape to allow easy loop access for crystal manipulation such as cryosoaking and rinsing. Plate is constructed of optically clear polystyrene to enable precise crystal visualization. Can be used with manual pipettors or automated liquid handling systems. Stackable plate is resistant to organic solvents, features reduced polarization, and may be bar coded
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty