Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Super-Nuova™ Multi-Position Digital Stirrers, 240 V

Supplier: Thermo Scientific
S136030-33Q
80089-838EA 2704.58 USD InStock
80089-838
Super-Nuova™ Multi-Position Digital Stirrers, 240 V
Stirrers Magnetic Stirrers, Multi-position
Super-Nuova™ Multi-Position Digital Stirrers offer four individually controlled stirring positions, ceramic tops and a 4-function memory.

  • 240 V model
  • Microprocessor control ensures accurate and stable setpoints for stirring speed
  • Stir Trac braking feature brings stir bar to an immediate stop for quick flask removal
  • Digital rpm display and stirring adjustable in units of 1 rpm
  • Single control knob controls speed; lock feature prevents accidental changes
  • RS-232 port outputs time count, stir speed (set point) directly to your PC
  • Seamless, reflective white ceramic surface aids sample visibility
  • Clean easily and resist alkalis and acids

Stirrers are designed for stronger coupling and stirring control with advanced safety features. Ideal for labs with multiple users. Rugged low-profile cast-aluminum body provides stability, durability and protects internal components from accidental spills.

Ideal for carrying out digestion, titration, and solubility and enzymatic studies.

Delivery information: Includes detachable line cord and plug.

Caution: 240 V model.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty